Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Mercury(II) chloride, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1080814

Quantity 25g

 View more versions of this product

Catalog No. AA1080814

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications Description & Specifications

Chemical Identifiers

7487-94-7
271.5
UN1624
Mercury(II) chloride
HgCl2
MFCD00011041
14,5876
Puratronic™

Specifications

7487-94-7
HgCl2
MFCD00011041
14,5876
Odorless
302°C
280° to 282°C
Mercury(II) chloride
Puratronic™
Powder
271.5
UN1624
5.4g/mL
1.8590
White
25g
99.999%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H372-H341-H361-H314
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURIC CHLORIDE

RTECSNumber : OV9100000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit