Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mercury(II) cyanide, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | White |
Physical Form | Fine Crystalline Powder |
Quantity | 5 g |
Assay Percent Range | 99% |
Linear Formula | Hg(CN)2 |
Fieser | 01,655; 03,197 |
Merck Index | 15,5944 |
Solubility Information | Solubility in water: 93g/L in water (14°C). Other solubilities: 1g/3mL in boiling water, 1g/13mL in alcohol, 1g/4mL in methanol, slowly soluble in glycerol, slightly soluble in ether |
Formula Weight | 252.63 |
Percent Purity | 99% |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement: Fatal if swallowed. Fatal in contact with skin. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. Fatal if inhaled. Contact with acids liberates very toxic gas. |
Precautionary Statement | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. Remove/Take off immediately all contaminated clothing. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 209-741-6 |
RTECSNumber | OW1515000 |
TSCA | TSCA |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction