Alfa Aesar™ Mercury(II) fluoride, 95%

Manufacturer: Alfa Aesar™ 1153309

Quantity 10g

 View more versions of this product

Catalog No. AA1153309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7783-39-3
    Linear Formula HgF2
    Formula Weight 238.61
    MDL Number MFCD00016139
    UN Number UN2025
    Merck Index 14,5878
    Chemical Name or Material Mercury(II) fluoride

    Specifications

    Formula Weight 238.61
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −10 Mesh
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Odor Odorless
    MDL Number MFCD00016139
    CAS 7783-39-3
    Color Yellow
    Melting Point 645°C (dec.)
    Density 8.95g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
    TSCA TSCA
    Physical Form Crystalline
    UN Number UN2025
    Merck Index 14,5878
    Health Hazard 1 Danger
    Sensitivity Moisture Sensitive
    Linear Formula HgF2
    Chemical Name or Material Mercury(II) fluoride
    Assay 95%

    GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit