Mercury(II) oxide, 99.998% (metals basis), Alfa Aesar™ Puratronic™

Manufacturer: Alfa Aesar 1081006

Quantity 5g

 View more versions of this product

Catalog No. AA1081006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21908-53-2
    Physical Form Powder
    Assay Percent Range 99.998% (metals basis)
    Molecular Formula HgO
    Formula Weight 216.59
    MDL Number MFCD00011045
    Chemical Name or Material Mercury(II) oxide

    Specifications

    Formula Weight 216.59
    Quantity 5g
    Color Dark orange
    DOT Information DOT Class: 6.1, Packaging Group: II
    EINECS Number 244-654-7
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.998% (metals basis)
    Molecular Formula HgO
    Appearance Dark orange powder
    MDL Number MFCD00011045
    CAS 21908-53-2
    Health Hazard 1 H300-H310-H330-H373-H400-H410: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Mercury(II) oxide

    H300-H310-H330-H373-H400-H410: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit