Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) oxide, 99.998% (metals basis), Alfa Aesar™, Puratronic™

Click to view available options
Quantity:
5g
25g
21908-53-2
HgO
MFCD00011045
mercuric oxide,mercury ii oxide,mercury oxide,mercuric oxide, red,red mercuric oxide,mercuric oxide, yellow,santar,red precipitate,mercury monoxide
21908-53-2
HgO
MFCD00011045
mercuric oxide,mercury ii oxide,mercury oxide,mercuric oxide, red,red mercuric oxide,mercuric oxide, yellow,santar,red precipitate,mercury monoxide
Assay Percent Range 99.998% (metals basis)
Appearance Dark orange powder
Solubility Information Partially soluble in water. insoluble in alcohol,ether,acetone and ammonia.
Formula Weight 216.59
Physical Form Powder
Density 11.14
Odor Odorless
Color Orange
Melting Point 500°C (decomposition)
Quantity 25g
Chemical Name or Material Mercury(II) oxide
Show More Show Less

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c

H300+H310+H330-H373

DOTInformation : DOT Class: 6.1, Packaging Group: II

EINECSNumber : 244-654-7

RTECSNumber : OW8750000

TSCA : Yes

Recommended Storage : Ambient temperatures