Alfa Aesar™ Mercury(II) perchlorate hexahydrate, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4431709

Quantity 10g

 View more versions of this product

Catalog No. AA4431709

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7616-83-3
    Molecular Formula Cl2HgO8·6H2O
    Linear Formula Hg(ClO4)2·6H2O
    Formula Weight 507.58 (399.49 Anhydrous)
    MDL Number MFCD00011036
    UN Number UN3087
    Chemical Name or Material Mercury (II) perchlorate hexahydrate

    Specifications

    Formula Weight 507.58 (399.49 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P283-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H271-H300-H310-H330-H373
    May cause fire or explosion
    strong oxidizer.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Physical Form Crystalline
    Molecular Formula Cl2HgO8·6H2O
    CAS 7616-83-3
    Color Off-white
    Quantity 10g
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
    TSCA TSCA
    Linear Formula Hg(ClO4)2·6H2O
    MDL Number MFCD00011036
    UN Number UN3087
    Health Hazard 1 Danger
    Chemical Name or Material Mercury (II) perchlorate hexahydrate
    Assay 99.999%, (metals basis)

    GHS H Statement
    H271-H300-H310-H330-H373
    May cause fire or explosion
    strong oxidizer.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P221-P283-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit