Learn More
Alfa Aesar™ Mercury(II) perchlorate trihydrate, Reagent Grade
Manufacturer:  Alfa Aesar™ 1163414
Quantity 25g
 View more versions of this product
Catalog No. AA1163414
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
73491-34-6
453.54 (399.49 Anhydrous)
UN3087
Reagent Grade
Hg(ClO4)2·3H2O
MFCD00149735
Mercury(II) perchlorate trihydrate
Specifications
73491-34-6
Hg(ClO4)2·3H2O
MFCD00149735
White
Mercury(II) perchlorate trihydrate
Crystalline
453.54 (399.49 Anhydrous)
UN3087
25g
Reagent Grade
Safety and Handling
SDS & Safety Documents
GHS H Statement
H271-H300-H310-H330-H373
May cause fire or explosion
strong oxidizer.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P221-P283-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
TSCA : TSCA