Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Mercury(II) selenide, 99.999% (metals basis)

Click to view available options
Quantity:
1g
5g
20601-83-6
HgSe
279.563
MFCD00049743
YQMLDSWXEQOSPP-UHFFFAOYSA-N
mercury selenide,mercury ii selenide,mercury-selenium complex,mercury monoselenide,mercury 2+ selenide,mercury ii selenide trace metals basis 1g
88609
selanylidenemercury
[Se]=[Hg]
20601-83-6
HgSe
279.563
MFCD00049743
YQMLDSWXEQOSPP-UHFFFAOYSA-N
mercury selenide,mercury ii selenide,mercury-selenium complex,mercury monoselenide,mercury 2+ selenide,mercury ii selenide trace metals basis 1g
88609
selanylidenemercury
[Se]=[Hg]
UN Number UN2025
Solubility Information Insoluble in water.
Formula Weight 279.55
Physical Form -100 Mesh Powder
Percent Purity 99.999%
Density 8.21 g/mL
Mesh Size −100 Mesh
Odor Odorless
Color Gray
Melting Point (sublimation)
Quantity 5g
Chemical Name or Material Mercury(II) selenide
Assay (metals basis)
Show More Show Less

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c

H301-H330-H360Df-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.

EINECSNumber : 243-910-5

TSCA : Yes

Recommended Storage : Ambient temperatures