Learn More
Alfa Aesar™ Mercury(II) silver iodide
Manufacturer:  Alfa Aesar™ 3930622
Quantity 100g
 View more versions of this product
Catalog No. AA3930622
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
7784-03-4
MFCD00046160
Mercury(II) silver iodide
923.95
UN2025
Specifications
7784-03-4
923.95
UN2025
Yellow
Mercury(II) silver iodide
Powder
MFCD00046160
Light Sensitive
100g
Safety and Handling
SDS & Safety Documents
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.