Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) sulfate, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
7783-35-9
HgO4S
296.648
DOBUSJIVSSJEDA-UHFFFAOYSA-L
mercury ii sulfate,mercuric sulfate,mercury sulphate,mercury bisulfate,mercury disulfate,mercury persulfate,mercuric sulphate,mercury sulfate hgso4,unii-j4l3ppg58i,mercuric bisulphate
24544
mercury(2+);sulfate
[O-]S(=O)(=O)[O-].[Hg+2]
7783-35-9
HgO4S
296.648
DOBUSJIVSSJEDA-UHFFFAOYSA-L
mercury ii sulfate,mercuric sulfate,mercury sulphate,mercury bisulfate,mercury disulfate,mercury persulfate,mercuric sulphate,mercury sulfate hgso4,unii-j4l3ppg58i,mercuric bisulphate
24544
mercury(2+);sulfate
[O-]S(=O)(=O)[O-].[Hg+2]
Assay Percent Range 99+%
Linear Formula HgSO4
Formula Weight 296.64
Physical Form Crystalline Powder
Percent Purity ≥99.0%
Density 6.4700g/mL
Packaging Glass Bottle
Color White
Quantity 5g
Chemical Name or Material Mercury(II) sulfate