Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mercury(Ii) Sulfate, Honeywell Fluka™
Click to view available options
Quantity:
250g
1kg
6 x 250g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Linear Formula | HgSO4 |
UN Number | 1645 |
Formula Weight | 296.65g/mol |
Percent Purity | ≥98% |
Grade | puriss. |
Quantity | 1kg |
Chemical Name or Material | Mercury(Ii) Sulfate |
Safety and Handling
P260-P262-P280-P302 + P352 + P310-P304 + P340 + P310
H300 + H310 + H330-H373-H410
EINECSNumber : 231-992-5
RTECSNumber : OX0500000