Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(Ii) Sulfate, Honeywell Fluka™

Click to view available options
Quantity:
250g
1kg
6 x 250g
7783-35-9
HgO4S
296.648
MFCD00011047
DOBUSJIVSSJEDA-UHFFFAOYSA-L
mercury ii sulfate,mercuric sulfate,mercury sulphate,mercury bisulfate,mercury disulfate,mercury persulfate,mercuric sulphate,mercury sulfate hgso4,unii-j4l3ppg58i,mercuric bisulphate
24544
mercury(2+);sulfate
[O-]S(=O)(=O)[O-].[Hg+2]
7783-35-9
HgO4S
296.648
MFCD00011047
DOBUSJIVSSJEDA-UHFFFAOYSA-L
mercury ii sulfate,mercuric sulfate,mercury sulphate,mercury bisulfate,mercury disulfate,mercury persulfate,mercuric sulphate,mercury sulfate hgso4,unii-j4l3ppg58i,mercuric bisulphate
24544
mercury(2+);sulfate
[O-]S(=O)(=O)[O-].[Hg+2]

Puriss.,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula HgSO4
UN Number 1645
Formula Weight 296.65g/mol
Percent Purity ≥98%
Grade puriss.
Quantity 250g
Chemical Name or Material Mercury(Ii) Sulfate

P260-P262-P280-P302 + P352 + P310-P304 + P340 + P310

H300 + H310 + H330-H373-H410

EINECSNumber : 231-992-5

RTECSNumber : OX0500000