Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mercury(II) thiocyanate, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 165.0°C |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 50 g |
Assay Percent Range | 99% min. (Complexometry) |
Linear Formula | Hg(SCN)2 |
Flash Point | 120°C |
Fieser | 09,293 |
Merck Index | 15, 5958 |
Solubility Information | Solubility in water: 0.7g/L (25°C). Other solubilities: soluble in diluted hcl |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Fatal if inhaled. Fatal if swallowed. Fatal in contact with skin. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement Rinse mouth. Wear protective gloves/protective clothing. IF ON SKIN: Gently wash with plenty of soap and water. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at r |
EINECSNumber | 209-773- |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction