Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury, Redistilled , Macron Fine Chemicals™

Click to view available options
Quantity:
1lb.
Packaging:
Poison Pack
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
7439-97-6
Hg
200.592
QSHDDOUJBYECFT-UHFFFAOYSA-N
hydrargyrum,quicksilver,metallic,liquid silver,mercurio,quecksilber,mercure,quick silver,colloidal,mercury, metallic
23931
CHEBI:16170
mercury
[Hg]
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
7439-97-6
Hg
200.592
QSHDDOUJBYECFT-UHFFFAOYSA-N
hydrargyrum,quicksilver,metallic,liquid silver,mercurio,quecksilber,mercure,quick silver,colloidal,mercury, metallic
23931
CHEBI:16170
mercury
[Hg]
Quantity 1lb.
Packaging Poison Pack
Formula Weight 200.59
Chemical Name or Material Mercury, Redistilled, Purified