Alfa Aesar™ Mercury tetrathiocyanatocobaltate(II)

Manufacturer: Alfa Aesar™ 1375914

Quantity 25g

 View more versions of this product

Catalog No. AA1375914

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 27685-51-4
    Linear Formula HgCo(CNS)4
    Formula Weight 491.83
    MDL Number MFCD00011048
    UN Number UN2025
    Synonym Cobalt(II) tetrathiocyanatomercurate
    Chemical Name or Material Mercury tetrathiocyanatocobaltate(II)

    Specifications

    Formula Weight 491.83
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H334-H373-H317
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN2025
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
    Physical Form Crystalline
    CAS 27685-51-4
    Synonym Cobalt(II) tetrathiocyanatomercurate
    Health Hazard 1 Danger
    TSCA TSCA
    Linear Formula HgCo(CNS)4
    MDL Number MFCD00011048
    Color Pale blue
    Quantity 25g
    Chemical Name or Material Mercury tetrathiocyanatocobaltate(II)

    GHS H Statement
    H300-H310-H330-H334-H373-H317
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit