Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Mercury tetrathiocyanatocobaltate(II)

Manufacturer:  Alfa Aesar™ 1375906

Quantity 5g

 View more versions of this product

Catalog No. AA1375906

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

27685-51-4
491.83
UN2025
Mercury tetrathiocyanatocobaltate(II)
HgCo(CNS)4
MFCD00011048
Cobalt(II) tetrathiocyanatomercurate

Specifications

27685-51-4
HgCo(CNS)4
MFCD00011048
Cobalt(II) tetrathiocyanatomercurate
5g
Crystalline
491.83
UN2025
Pale blue
Mercury tetrathiocyanatocobaltate(II)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H310-H330-H334-H373-H317
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit