Meridian Bioscience™ illumigene™ Mycoplasma DNA Amplification Assay Kit and Accessories
Provides rapid diagnosis, leading to earlier detection of outbreaks
Manufacturer: MERIDIAN BIOSCIENCE 610172MYCO
 View more versions of this product
Catalog No. 23-029-260
  Check Availability
Add to Cart
    Specifications
    Product Type Mycoplasma Starter Package
    CPT Code 87581
    Detectable Analytes Mycoplasma DNA in Human throat and nasopharyngeal swabs
    Includes Phosphate-buffered solution (assay control II), Tris-buffered solution (reaction Buffer II), mycoplasma test device, screw-top microcentrifuge tubes
    Quantity Each
    Sample Type Throat and nasopharyngeal swab specimens (0.85% Saline, M4, M4-RT, M5, UTM-RT)
  • Description & Specifications
Related Products
Product Content Feedback