Available on GSA/VA Contract for Federal Government customers only. Meridian Bioscience™ illumigene External Control Kit
for the detection of C. difficiles in stool
illumigene C. difficile External Control Reagents are supplied separately It is recommended that the reactivity of each new lot and each new shipment of illumigene C. difficile be verified on receipt and before use. External control tests should be performed thereafter in accordance with appropriate federal, state and local guidelines. The illumigene C. difficile test kit should not be used in patient testing if the external controls do not produce the correct results.
Online instant savings
$214.25
$238.05
  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
23-029-253
Available on GSA/VA Contract for Federal Government customers only. 
Meridian Bioscience
279920
External Control Kit
Each for $214.25
Each for $238.05
Online Instant Savings (10%)
Check Availability
Add to Cart
Descriptions & Specifications
Related Products