Learn More
Meridian Bioscience™ illumigene™ Bordetella Pertussis Kit
Diagnostic system for detection of Bordetella pertussis in nasopharyngeal swab samples
Manufacturer:  MERIDIAN BIOSCIENCE 280750
Catalog No. 23-029-263
Due to product restrictions, please to purchase or view availability for this product.
Includes: illumigene™ Pertussis Assay Control/Negative Control Reagent, illumigene Pertussis Test Device, illumigene Pertussis Sample Buffer, Mineral Oil
Specifications
Description
Bordetella Pertussis is the pathogen responsible for the endemic respiratory disease whooping cough. Although there is a vaccine available, it is currently showing an increase of reportable infections in the US. Molecular assays are increasingly being used due to improved sensitivity and rapid turnaround time.
  • Based on loop-mediated isothermal DNA amplification (LAMP) technology
  • Result interpretation in less than 1 hour by illumipro-10 Incubator/Reader
  • High sensitivity (87.8%) and specificity (97.8%)
  • Quicker initiation of patient treatment
  • Small instrument footprint
  • Simple testing protocol, easily train all laboratory personnel
The device is not intended for point-of-care use.
Healthcare industry
Description & Specifications
Specifications
Detection of Bordetella pertussis in nasopharyngeal swab samples
50 Tests
87.8%
2° to 30°C
Safety and Handling
ShelfLife : 18 months