Meridian Bioscience™ illumigene™ Bordetella Pertussis Starter Pack

Diagnostic system for detection of Bordetella pertussis in nasopharyngeal swab samples

Manufacturer: MERIDIAN BIOSCIENCE 610172BP

Catalog No. 23-029-265

  Check Availability
Add to Cart

Includes: illumigene™ Pertussis Assay Control/Negative Control Reagent, illumigene Pertussis Test Device, illumigene Pertussis Sample Buffer, Mineral Oil

    Specifications

    Storage Requirements 2° to 30°C
    For Use With (Application) Detection of Bordetella pertussis in nasopharyngeal swab samples
    Quantity 50 Tests
    Sensitivity Drift 87.8%
    Shelf Life 18 months

    Bordetella Pertussis is the pathogen responsible for the endemic respiratory disease whooping cough. Although there is a vaccine available, it is currently showing an increase of reportable infections in the US. Molecular assays are increasingly being used due to improved sensitivity and rapid turnaround time.

    • Based on loop-mediated isothermal DNA amplification (LAMP) technology
    • Result interpretation in less than 1 hour by illumipro-10 Incubator/Reader
    • High sensitivity (87.8%) and specificity (97.8%)
    • Quicker initiation of patient treatment
    • Small instrument footprint
    • Simple testing protocol, easily train all laboratory personnel

    The device is not intended for point-of-care use.

    Healthcare industry

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit