Learn More
Please call 1-800-766-7000 to place your order or try our site again later.
Meridian Bioscience Para-Pak CON-Trate Concentration Systems
Injury-free parasite retrieval
$1189.00
Specifications
Certifications/Compliance | CE |
---|---|
Storage Requirements | 20 to 30°C |
Includes: Caps, filtration devices, 15mL graduated centrifuge tubes, CON-Trate Reagent A (MucoPenX) and Reagent B (ethyl acetate)
Catalog Number | Mfr. No. | Product Type | Quantity | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Type | Price | Quantity & Availability | |||
14-910-165
![]() |
Meridian Bioscience
960200 |
Stool Concentration System | 200 Case | Preserved and unpreserved stool |
Case of 200 for $1,189.00
|
N/A |
Description
- Designed to optimize parasitic concentration/recovery and minimize specimen preparation time
- Kits of 50 or 500 tests
- Filtration devices also available separately
Order Info
Packaged in a workstation design to reduce procedure time
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
CE | |
20 to 30°C |
Safety and Handling
ShelfLife : 3 Year(s)
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.