Learn More
Meridian Bioscience™ Para-Pak™ CON-Trate™ Concentration Systems
Injury-free parasite retrieval
Manufacturer:  MERIDIAN BIOSCIENCE 960500
 View more versions of this product
Catalog No. 23-312020
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CE
Stool Concentration System
Workstation
500 Test(s)
Cap, filtration devices, 15mL graduated centrifuge tubes, Con-Trate™ reagent A (MucoPenX) and reagent B (Ethyl Acetate)
20 to 30°C
500 Case
Preserved and unpreserved stool
Safety and Handling
ShelfLife : 3 Year(s)