Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Para-Pak SpinCon™ Concentration System

Requires only one spin for recovery of protozoan cysts, trophs and helminth ova and larvae

$865.00 - $3003.00

Specifications

Format Tube
Sample Type Stool
Type One-spin Specimen Concentration System
For Use With (Application) Concentrates eggs, larvae and protozoa from preserved stools
Includes Spincon devices, 15mL tubes, funnels, caps and surfactant
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-315-085A
View Documents
Meridian Bioscience™
972200
200 Case
Case of 200 for $865.00
Edge
Add to cart
 
23-315-085B
View Documents
Meridian Bioscience™
972500
500 Case
Case of 200 for $3,003.00
Add to cart
 
Description

Description

  • Ethyl acetate-free
  • Based on filtration requiring no chemicals
  • Contains Spincon devices, funnels, caps 15mL tubes and surfactant
  • Stool specimen
  • Shelf life of 36 months

Certifications

CE marked

Specifications

Specifications

Tube
One-spin Specimen Concentration System
Spincon devices, 15mL tubes, funnels, caps and surfactant
Stool
Concentrates eggs, larvae and protozoa from preserved stools
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit