Learn More
Meridian Bioscience™ MERIFLUOR™ EBV VCA IgG Immunofluorescence Test
SDP
An immunofluorescence test for the detection of IgG Antibodies to Epstein-Barr Virus
Supplier:  Meridian Bioscience™ EB100
Catalog No. 23-043-155
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. View substitute products »
Alert:
The conjugate contains Evans Blue dye as a counterstain. This dye may be carcinogenic. Avoid contact with the skin.
The PBS Powder contains sodium azide, which may react with lead or copper plumbing to form highly explosive metal azide compounds. When disposing of reagents through plumbing fixtures, flush with a large volume of water to prevent azide build-up in drains.
Sodium azide is very toxic in case of ingestion. In contact with acid it emits a very toxic gas. In case of contact with the skin wash immediately and abundantly with water.
Control sera are considered POTENTIALLY BIOHAZARDOUS MATERIAL. Each donor unit used in the preparation of the control sera was tested by an FDA-cleared method for the presence of HBsAg, antibodies to HIV-1, HIV-2 and HCV and found to be negative. However, because no test method can offer complete assurance that infectious agents are absent, sera and all patients' specimens should be handled at the Biosafety Level 2.
This item is not returnable. View return policy
Description
Description
  • The indirect fluorescent antibody (IFA) method is used in the MERIFLUOR EBV VCA IgG IFA Test
  • Lymphocytes from a Burkitt's patient are used as the VCA substrate
  • Patient serum is reacted with the substrate and if antibodies to EBV are present they will bind to the antigen substrate and not rinse off When fluoresceinated antihuman IgG is added to the reaction site, it will bind to the IgG antibodies, causing the EBV-infected cells to fluoresce when viewed through the fluorescenc microscope
  • For in vitro diagnostic use
  • Kit includes EBV Antigen Substrate Slides, Phosphate Buffered Saline (PBS) Powder, Positive and Negative Control Sera, Conjugate and Mounting Fluid
Specifications
Specifications
CE Marked
IFA
Epstein-Barr Virus (EBV) IgG IFA Test Kit
EBV Antigen substrate slides, Phosphate Buffered Saline powder, positive and negative control sera, conjugate and mounting fluid
EBV-VCA IgG
Serum
An immunofluorescence test for the detection of IgG Antibodies to Epstein-Barr Virus
Each