Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MERIT40/HSPC142 Mouse anti-Human, Alexa Fluor™ 594, Clone: 1251, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243621AF594

Catalog No. NP243621A94


Add to cart

Description

Description

MERIT40/HSPC142 Monoclonal antibody specifically detects MERIT40/HSPC142 in Human samples. It is validated for Western Blot,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

MERIT40/HSPC142
1251
Western Blot, Immunocytochemistry/Immunofluorescence
BRISC and BRCA1 A complex member 1, C10orf62, C19orf62, chromosome 19 open reading frame 62, FLJ20571, HSPC142, mediator of Rap80 interactions and targeting 40 kDa, Mediator of RAP80 interactions and targeting subunit of 40 kDa, MERIT40BRCA1-A complex subunit MERIT40, NBA1BRISC and BRCA1 A complex member, new component of the BRCA1 A complex, New component of the BRCA1-A complex, new component of the BRCAA1 A complex
Mouse
IgG2a
Store at 4°C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
BABAM1
The exact sequence is proprietary.
0.1 mL
Primary
29086
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit