Learn More
Beaver Visitec Intl LtdSupplier Diversity Partner MEROCEL EYE SPEAR,180 COUNT, 10 SPEARS/POUCH DFS Item
Manufacturer:  Beaver Visitec Intl LtdSupplier Diversity Partner 400101
Made from exclusive Merocel polyvinyl acetal (PVA) material. The Merocel material is highly absorbent, fast wicking and lint free. Designed to provide the surgeon with superior ber free products for unparalleled uid control. Blue handle. 7.16 cm (2.82 in) overall length.180 count, 10 per pouch, 18 pouches per box
Catalog No. NC0093269