Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Merrifield polymer resin, 1%crossl. with DVB, 2.5-3.0 mmol Cl/g,200-400 mesh, Acros Organics

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
9036-15-1
C19H19Cl
282.811
MFCD00146406
YDYHOUQBBDDITA-UHFFFAOYSA-N
Chloromethylated styrene-divinylbenzene copolymer
165755
1,2-bis(ethenyl)benzene;1-(chloromethyl)-2-ethenylbenzene
C=CC1=CC=CC=C1CCl.C=CC1=CC=CC=C1C=C
9036-15-1
C19H19Cl
282.811
MFCD00146406
YDYHOUQBBDDITA-UHFFFAOYSA-N
Chloromethylated styrene-divinylbenzene copolymer
165755
1,2-bis(ethenyl)benzene;1-(chloromethyl)-2-ethenylbenzene
C=CC1=CC=CC=C1CCl.C=CC1=CC=CC=C1C=C
For solid support synthesis
Additional Information crosslinking: 1% DVB
Loading 2.5 to 3mmol/g
Mesh Size 200 to 400 mesh
Packaging Glass bottle
Quantity 10g
Color White to Yellow
Physical Form Beads
Chemical Name or Material Merrifield polymer resin