Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MERS Coronavirus NP Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA5120759

Encompass

Catalog No. PIPA5120759


Add to Cart
This item is not returnable. View return policy

Description

Description

MERS Coronavirus NP Polyclonal antibody specifically detects MERS Coronavirus NP in Virus samples. It is validated for Western Blot
Specifications

Specifications

MERS Coronavirus NP
Polyclonal
Unconjugated
Coronavirus; MERS; NP
Recombinant protein of MERS-CoV Nucleoprotein.
100 μL
Primary
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.799 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Rabbit
Antigen affinity chromatography
RUO
Virus
Antibody
IgG
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit