Learn More
ZeptoMetrix MERS-CoV (Strain: Florida/USA-2_Saudi Arabia_2014) Culture Fluid
MERS-CoV (Strain: Florida/USA-2_Saudi Arabia_2014) Culture Fluid is an enveloped, positive-sense single-stranded RNA virus from the Coronaviridae family.
Manufacturer:  ZeptoMetrix 0810575CFHI
Catalog No. 22-158-341
Add to cart
Description
Description
  • Heat inactivated viral culture fluid sold as consumable testing materials
  • The suitability and performance characteristics should be determined by your laboratory for each intended usage
  • Viral inactivation is verified after heat inactivation by the absence of viral growth in tissue culture-based infectivity assays
  • Standard/Control
Specifications
Specifications
Repetitive freezing and thawing is not recommended (aliquot material if necessary). Titer will be altered by multiple freeze-thaws.
Each frozen aliquot contains 1 mL of heat inactivated viral culture fluid.
Viral culture fluids should be stored at -65°C or below.
Culture Fluid
Florida/USA-2_Saudi Arabia_2014
To avoid cross-contamination, use separate pipette tips for all reagents.
1 mL