Learn More
ZeptoMetrix MERS-CoV (Strain: Florida/USA-2_Saudi Arabia_2014) Lysate
MERS-CoV (Strain: Florida/USA-2_Saudi Arabia_2014) is an enveloped, positive-sense single-stranded RNA virus from the Coronaviridae family.
$673.00 - $1325.00
Specifications
Description For research or further manufacturing only. Not for use in diagnostic procedures
Storage Requirements Recommended storage temperature -65°C or below
Type Lysate
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
22-158-380
View Documents
ZeptoMetrix
08105750.5MG
0.5 mg
Each for $673.00
Add to cart
22-158-379
View Documents
ZeptoMetrix
0810575
1 mg
Each for $1,325.00
Add to cart
Description
Description
  • This product is intended for research, product development, quality assurance testing, or further manufacturing use.
  • Viral lysates can be used as an antigen, as a source for the purification of viral proteins, or in assays for the detection of viral antibodies.
Specifications
Specifications
For research or further manufacturing only. Not for use in diagnostic procedures
Recommended storage temperature -65°C or below
Lysate