Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ MES, 0.2M buffer soln., pH 7.0, low endotoxin

Click to view available options
Quantity:
100mL
250mL
145224-94-8
C6H15NO5S
213.248
MFCD00283848
MIIIXQJBDGSIKL-UHFFFAOYSA-N
16218417
2-morpholin-4-ylethanesulfonic acid;hydrate
C1COCCN1CCS(=O)(=O)O.O
145224-94-8
C6H15NO5S
213.248
MFCD00283848
MIIIXQJBDGSIKL-UHFFFAOYSA-N
16218417
2-morpholin-4-ylethanesulfonic acid;hydrate
C1COCCN1CCS(=O)(=O)O.O
Concentration 0.2M buffer soln.
Physical Form Liquid
pH 7.0
Color Colorless
Quantity 250mL
Chemical Name or Material MES, low endotoxin

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA