Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ MES, 0.2M buffer soln., pH 7.4, low endotoxin

Click to view available options
Quantity:
100mL
250mL
145224-94-8
C6H15NO5S
213.248
MFCD00283848
MIIIXQJBDGSIKL-UHFFFAOYSA-N
16218417
2-morpholin-4-ylethanesulfonic acid;hydrate
C1COCCN1CCS(=O)(=O)O.O
145224-94-8
C6H15NO5S
213.248
MFCD00283848
MIIIXQJBDGSIKL-UHFFFAOYSA-N
16218417
2-morpholin-4-ylethanesulfonic acid;hydrate
C1COCCN1CCS(=O)(=O)O.O
Concentration 0.2M buffer soln.
Physical Form Liquid
pH 7.4
Color Colorless
Quantity 250mL
Chemical Name or Material MES, low endotoxin