Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MES, Monohydrate, Free Acidline, Crystal, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
20g
200g
500g
Packaging:
Poly Bottle
POLYSTORMOR
145224-94-8
C6H15NO5S
213.248
MIIIXQJBDGSIKL-UHFFFAOYSA-N
mes monohydrate,2-morpholinoethanesulfonic acid hydrate,mes hydrate,mes buffer hydrate,4-morpholineethanesulfonic acid monohydrate,4-morpholineethanesulfonic acid, monohydrate,2-4-morpholinyl ethanesulfonic acid hydrate,mes hydrate buffer,4-morpholineethanesulfonicacid, hydrate 1:1,mesmonohydrate
16218417
2-morpholin-4-ylethanesulfonic acid;hydrate
C1COCCN1CCS(=O)(=O)O.O
145224-94-8
C6H15NO5S
213.248
MIIIXQJBDGSIKL-UHFFFAOYSA-N
mes monohydrate,2-morpholinoethanesulfonic acid hydrate,mes hydrate,mes buffer hydrate,4-morpholineethanesulfonic acid monohydrate,4-morpholineethanesulfonic acid, monohydrate,2-4-morpholinyl ethanesulfonic acid hydrate,mes hydrate buffer,4-morpholineethanesulfonicacid, hydrate 1:1,mesmonohydrate
16218417
2-morpholin-4-ylethanesulfonic acid;hydrate
C1COCCN1CCS(=O)(=O)O.O

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology Applications
Quantity 20g
Packaging POLYSTORMOR
Formula Weight 213.25
Physical Form Crystals
Grade Ultrapure Bioreagent
Chemical Name or Material MES, Monohydrate, Free Acidline