Learn More
Apexbio Technology LLC Mesalamine 89-57-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19695.1
Encompass_Preferred
Mesalamine (89-57-6) is a small-molecule inhibitor targeting the I B kinase (IKK) signaling pathway It is designed to modulate the inflammatory response thereby regulating the NF- B pathway and the expression of pro-inflammatory mediators Mesalamine exerts its biological activity primarily through inhibition of cytokine-mediated activation of the NF- B pathway Based on these pharmacological properties mesalamine holds research potential in studies of inflammatory signaling cascades autoimmune conditions and related inflammatory disorders
Catalog No. 50-000-93049