Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mesalamine, USP, 98.5-101.5%, Spectrum™ Chemical

Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Mesalamine, USP is used primarily as an anti-inflammatory drug in the treatment of ulcerative colitis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Ignition Residue | 0.2% |
Loss on Drying | 0.5% |
Packaging | Amber Glass Bottle |
Quantity | 25 g |
Percent Purity | 98.5 to 101.5% |
Grade | USP |