Learn More
Meshing Around with Gears Kit Products for Science Education
Students will be well on their way to understanding how gears work!
$1.61 - $13.52
Alert:
CHOKING HAZARD—Small parts. Not for children under 3 yr.
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S94409 Products for Science Education
View Documents
Si Metric Manufacturing Limited
70021
Meshing Around With Gears Kit
Each for $13.52
Add to cart
S93922 Products for Science Education
View Documents
Si Metric Manufacturing Limited
21801
Teacher'S Guide
Each for $1.61
Add to cart
Description
Description
  • Comprehensive kit with all of the materials required to build basic models
  • Models include a winch and a trundle cart using gears, corroplast and JAX clips
  • 16-page teacher's guide covers many basic gearing principles including direction of rotation, idlers, gearing up and down, and compound gearing