Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Mesitylene-2-sulfonyl hydrazide, 97%

Manufacturer:  Alfa Aesar™ H5558606

Quantity 5g

 View more versions of this product

Catalog No. AAH5558606


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

16182-15-3
MFCD00007585
2118380
Mesitylene-2-sulfonyl hydrazide
214.28
UN1325
2,4,6-Trimethylbenzenesulfonyl hydrazide

Specifications

16182-15-3
214.28
UN1325
2,4,6-Trimethylbenzenesulfonyl hydrazide
112° to 114°C (dec.)
Mesitylene-2-sulfonyl hydrazide
Powder
MFCD00007585
2118380
White to off-white
5g
97%

Safety and Handling

SDS & Safety Documents

DOTInformation : Transport Hazard Class: 4.1; Packaging Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit