Alfa Aesar™ Mesitylene, 98+%

Manufacturer: Alfa Aesar™ A11323AP

Quantity 500mL

 View more versions of this product

Catalog No. AAA11323AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-67-8
    Molecular Formula C9H12
    Formula Weight 120.2
    MDL Number MFCD00008538
    UN Number UN2325
    Beilstein 906806
    Merck Index 14,5907
    Synonym 1,3,5-Trimethylbenzene
    Chemical Name or Material Mesitylene

    Specifications

    Formula Weight 120.2
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C9H12
    CAS 108-67-8
    Melting Point −45°C
    MDL Number MFCD00008538
    UN Number UN2325
    Synonym 1,3,5-Trimethylbenzene
    Boiling Point 163° to 165°C
    Health Hazard 2 GHS H Statement
    H226-H335
    Flammable liquid and vapor.
    May cause respiratory irritation.
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 1,3,5-TRIMETHYLBENZENE
    RTECS Number OX6825000
    TSCA TSCA
    Beilstein 906806
    Merck Index 14,5907
    Quantity 500mL
    Flash Point 44°C (111°F)
    Odor Unpleasant
    Chemical Name or Material Mesitylene
    Assay ≥98%

    Description

    Applications
    Mesitylene is used to make plastics and dyes. It acts as a solvent, ligand in organometallic chemistry and precursor to 2,4,6-trimethylaniline. It is also used as a developer for photopatternable silicones due to its solvent properties in the electronics industry. It is used as an internal standard in nuclear magnetic resonance (NMR) samples due to the presence of three equivalent protons in it. It is involved in the production of trimesic acid and antioxygen, epoxy firming agent and polyester resin stabilizers. Further, it is used as an additive and component in aviation gasoline blends.

    Solubility
    Partly miscible with water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H226-H335
    Flammable liquid and vapor.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit