Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesitylene, 99%, Extra Pure, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
108-67-8
C9H12
120.19
MFCD00008538
AUHZEENZYGFFBQ-UHFFFAOYSA-N
mesitylene,sym-trimethylbenzene,benzene, 1,3,5-trimethyl,fleet-x,trimethylbenzol,3,5-dimethyltoluene,s-trimethylbenzene,hsdb 92,2,4,6-trimethylbenzene,1,3,5-trimethyl-benzene
7947
CHEBI:34833
1,3,5-trimethylbenzene
CC1=CC(=CC(=C1)C)C
108-67-8
C9H12
120.19
MFCD00008538
AUHZEENZYGFFBQ-UHFFFAOYSA-N
mesitylene,sym-trimethylbenzene,benzene, 1,3,5-trimethyl,fleet-x,trimethylbenzol,3,5-dimethyltoluene,s-trimethylbenzene,hsdb 92,2,4,6-trimethylbenzene,1,3,5-trimethyl-benzene
7947
CHEBI:34833
1,3,5-trimethylbenzene
CC1=CC(=CC(=C1)C)C
Viscosity 0.834 mm2/s (20°C)
Boiling Point 163.0°C to 166.0°C
Refractive Index 1.4980 to 1.5000
Specific Gravity 0.8675
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1L
Flash Point 44°C
Melting Point -45.0°C
Density 0.8675g/mL
Color Undesignated
Assay Percent Range 99%
Linear Formula C6H3(CH3)3
Beilstein 05, 406
Merck Index 15, 5977
Solubility Information Solubility in water: 2.9g/L (20°C). Other solubilities: miscible with alcohol,benzene and ether
Formula Weight 120.19
Physical Form Liquid
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Mesitylene
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning