Alfa Aesar™ (Mesitylene)tungsten tricarbonyl, 98%

Manufacturer: Alfa Aesar™ 3958903

Quantity 1g

 View more versions of this product

Catalog No. AA3958903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12129-69-0
    Formula Weight 388.08
    MDL Number MFCD00015319
    UN Number UN3466
    Synonym Tricarbonyl(mesitylene)tungsten
    Chemical Name or Material (Mesitylene)tungsten tricarbonyl

    Specifications

    Formula Weight 388.08
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Tricarbonyl(mesitylene)tungsten
    Health Hazard 1 Danger
    Melting Point 165°C (dec.)
    Physical Form Crystalline
    CAS 12129-69-0
    UN Number UN3466
    Quantity 1g
    MDL Number MFCD00015319
    Health Hazard 2 GHS H Statement
    H300-H315-H319-H335
    Fatal if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
    Chemical Name or Material (Mesitylene)tungsten tricarbonyl
    Assay 98%

    GHS H Statement
    H300-H315-H319-H335
    Fatal if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit