Alfa Aesar™ Mesitylenesulfonic acid dihydrate, 98+%

Manufacturer: Alfa Aesar™ B2532122

Quantity 100g

 View more versions of this product

Catalog No. AAB2532122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 835617-36-2
    Molecular Formula C9H12O3S·2H2O
    Formula Weight 236.29 (200.26 Anhydrous)
    MDL Number MFCD00149535
    UN Number UN2585
    Beilstein 1961501
    Synonym 2,4,6-Trimethylbenzenesulfonic acid dihydrate
    Chemical Name or Material Mesitylenesulfonic acid dihydrate

    Specifications

    Formula Weight 236.29 (200.26 Anhydrous)
    Synonym 2,4,6-Trimethylbenzenesulfonic acid dihydrate
    Health Hazard 1 Danger
    Melting Point 74° to 77°C
    Quantity 100g
    Molecular Formula C9H12O3S·2H2O
    MDL Number MFCD00149535
    CAS 835617-36-2
    UN Number UN2585
    Beilstein 1961501
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
    Chemical Name or Material Mesitylenesulfonic acid dihydrate
    Assay ≥98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit