Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Mesitylenesulfonic acid sodium salt hemihydrate, 98%
Click to view available options
Quantity:
25g
100g
500g
Specifications
Quantity | 500g |
Melting Point | >300°C |
Beilstein | 5784990 |
Formula Weight | 231.26 (222.25 Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | Mesitylenesulfonic acid sodium salt hemihydrate |
Safety and Handling
EINECSNumber : 228-157-2
TSCA : Yes
Recommended Storage : Ambient temperatures