Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ meso-2,3-Dibromosuccinic acid, 98%

Click to view available options
Quantity:
100g
250g
500g
2500g
608-36-6
C4H4Br2O4
275.88
MFCD00066439
FJWGRXKOBIVTFA-XIXRPRMCSA-N
2r,3s-2,3-dibromosuccinic acid,meso-2,3-dibromosuccinic acid,unii-d2ii9ugq9x,d2ii9ugq9x,butanedioic acid, 2,3-dibromo-, 2r,3s-rel,2r,3s-rel-2,3-dibromosuccinic acid,2r,3s-2,3-dibromobutanedioic acid,meso-dibromosuccinic acid,2,3-dibromosuccinic acid, meso,2,3-dibromosuccinic acid meso-form mi
641611
(2S,3R)-2,3-dibromobutanedioic acid
C(C(C(=O)O)Br)(C(=O)O)Br
608-36-6
C4H4Br2O4
275.88
MFCD00066439
FJWGRXKOBIVTFA-XIXRPRMCSA-N
2r,3s-2,3-dibromosuccinic acid,meso-2,3-dibromosuccinic acid,unii-d2ii9ugq9x,d2ii9ugq9x,butanedioic acid, 2,3-dibromo-, 2r,3s-rel,2r,3s-rel-2,3-dibromosuccinic acid,2r,3s-2,3-dibromobutanedioic acid,meso-dibromosuccinic acid,2,3-dibromosuccinic acid, meso,2,3-dibromosuccinic acid meso-form mi
641611
(2S,3R)-2,3-dibromobutanedioic acid
C(C(C(=O)O)Br)(C(=O)O)Br
Applications
meso-2,3-dibromosuccinic acid is used in organic synthesis & pharmaceutical intermediate.

Solubility
Soluble in alcohol and ether. Slightly soluble in water and methenyl chloride.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, bases.
Melting Point ~280°C
Quantity 500g
UN Number UN3261
Beilstein 1725173
Merck Index 14,3029
Formula Weight 275.89
Percent Purity 98%
Chemical Name or Material meso-2,3-Dibromosuccinic acid

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.