Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-2,3-Dibromosuccinic acid, 98%, Thermo Scientific™

Catalog Number p-7027545
Click to view available options
Quantity:
100 g
250 g
500 g
2500 g
608-36-6
C4H2Br2O4
273.87
MFCD00066439
FJWGRXKOBIVTFA-XIXRPRMCSA-L
2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi
641611
(2S,3R)-2,3-dibromobutanedioic acid
[O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O
608-36-6
C4H2Br2O4
273.87
MFCD00066439
FJWGRXKOBIVTFA-XIXRPRMCSA-L
2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi
641611
(2S,3R)-2,3-dibromobutanedioic acid
[O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
meso-2,3-dibromosuccinic acid is used in organic synthesis & pharmaceutical intermediate.

Solubility
Soluble in alcohol and ether. Slightly soluble in water and methenyl chloride.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, bases.
TRUSTED_SUSTAINABILITY
Melting Point ∼280°C
Quantity 100 g
UN Number UN3261
Beilstein 1725173
Merck Index 14,3029
Solubility Information Soluble in alcohol and ether. Slightly soluble in water and methenyl chloride.
Formula Weight 275.89
Percent Purity 98%
Chemical Name or Material meso-2,3-Dibromosuccinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 208-396-9
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit