Alfa Aesar™ meso-2,3-Dibromosuccinic acid, 98%

Manufacturer: Alfa Aesar™ B2438230

Quantity 250g

 View more versions of this product

Catalog No. AAB2438230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 608-36-6
    Molecular Formula C4H4Br2O4
    Formula Weight 275.89
    MDL Number MFCD00066439
    UN Number UN3261
    Beilstein 1725173
    Merck Index 14,3029
    Chemical Name or Material meso-2,3-Dibromosuccinic acid

    Specifications

    Formula Weight 275.89
    MDL Number MFCD00066439
    Melting Point ≈280°C
    Quantity 250g
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Molecular Formula C4H4Br2O4
    CAS 608-36-6
    UN Number UN3261
    Beilstein 1725173
    Merck Index 14,3029
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material meso-2,3-Dibromosuccinic acid
    Assay 98%

    Description

    Applications
    meso-2,3-dibromosuccinic acid is used in organic synthesis & pharmaceutical intermediate.

    Solubility
    Soluble in alcohol and ether. Slightly soluble in water and methenyl chloride.

    Notes
    Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, bases.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit