Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ meso-Hydrobenzoin, 99%

Click to view available options
Quantity:
5g
25g
579-43-1
C14H14O2
214.264
MFCD00064253
IHPDTPWNFBQHEB-OKILXGFUSA-N
meso-hydrobenzoin,meso-1,2-diphenyl-1,2-ethanediol,unii-co9a49a84i,meso-stilbene glycol,1r,2s-1,2-diphenylethane-1,2-diol,meso-1,2-diphenylethylene glycol,hydrobenzoin, meso,unii-q61g3433lb component
853018
CHEBI:50015
(1R,2S)-1,2-diphenylethane-1,2-diol
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O
579-43-1
C14H14O2
214.264
MFCD00064253
IHPDTPWNFBQHEB-OKILXGFUSA-N
meso-hydrobenzoin,meso-1,2-diphenyl-1,2-ethanediol,unii-co9a49a84i,meso-stilbene glycol,1r,2s-1,2-diphenylethane-1,2-diol,meso-1,2-diphenylethylene glycol,hydrobenzoin, meso,unii-q61g3433lb component
853018
CHEBI:50015
(1R,2S)-1,2-diphenylethane-1,2-diol
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O
Applications
meso- Hydrobenzoin has been used in the preparation of trans-methyl meso- hydrobenzoin phosphite.

Solubility
Soluble in hot alcohol or chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Quantity 5g
Melting Point 134°C to 136°C
Beilstein 2050814
Merck Index 14,4777
Formula Weight 214.27
Percent Purity 99%
Chemical Name or Material meso-Hydrobenzoin