Learn More
meso-Tetraphenylporphine, low chlorine, Thermo Scientific Chemicals
Description
meso-Tetraphenylporphyrin is a porphyrin-based compound of interest in molecular electronics and supramolecular building blocks. Porphyrins are often used to construct structures in supramolecular chemistry. It finds it application as a sensitizer used for the photochemical generation of singlet oxygen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
meso-Tetraphenylporphyrin is a porphyrin-based compound of interest in molecular electronics and supramolecular building blocks. Porphyrins are often used to construct structures in supramolecular chemistry. It finds it application as a sensitizer used for the photochemical generation of singlet oxygen.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 449°C to 451°C |
Quantity | 1 g |
Beilstein | 379542 |
Solubility Information | Insoluble in water. |
IUPAC Name | 5,10,15,20-tetraphenyl-21,22-dihydroporphyrin |
Formula Weight | 614.71 |
Chemical Name or Material | meso-Tetraphenylporphine, low chlorine |
Safety and Handling
Hazard Category | H315-H319-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 213-025-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.