Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mesothelin, Mouse anti-Human, Biotin, Clone: TCL1/2079, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279834B
Description
Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Mesothelin | |
TCL1/2079 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
MSLN | |
Recombinant fragment (around aa 2-109) of human Mesothelin protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein | |
Mouse | |
IgG2a Kappa | |
0.1 mL | |
Cancer | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title