Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesothelin Monoclonal Antibody (C6), Invitrogen™

Mouse Monoclonal Antibody

$211.50 - $451.50

Specifications

Antigen Mesothelin
Clone C6
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA543664
Encompass
View Documents
Thermo Scientific
MA543664
20 μL
Each for $211.50
Add to Cart
 
PIMA543663
Encompass
View Documents
Thermo Scientific
MA543663
100 μL
Each for $451.50
Add to Cart
 
Description

Description

Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

Mesothelin
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300
CAK1; CAK1 antigen; megakaryocyte potentiating factor; Megakaryocyte-potentiating factor; Mes; Mesothelin; Mesothelin, cleaved form; MPF; MSLN; Pre-pro-megakaryocyte-potentiating factor; SMR; SMRP; soluble MPF mesothelin related protein
MSLN
IgG1 κ
Protein A/G
Antibody
C6
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human
Q13421
10232
Glu296-Ser606 (Accession Q13421), with N-terminal His Tag
Primary
-20°C
MSLN
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit