Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Mesothelioma Ab-1, Mouse Monoclonal Antibody

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Thermo Scientific™ Mesothelioma Ab-1, Mouse Monoclonal Antibody.

$369.50 - $498.00

Specifications

Antigen Mesothelioma Ab-1
Clone HBME-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS1494S1
View Documents
Thermo Scientific™
MS1494S1
500μL Each for $369.50
Add to cart
 
MS1494S
View Documents
Thermo Scientific™
MS1494S
1mL Each for $498.00
Add to cart
 
MS1494R7
View Documents
Thermo Scientific™
MS1494R7
7mL Each for $386.50
Add to cart
 
MS1494RQ
View Documents
Thermo Scientific™
MS1494RQ
12mL Each for $431.00
Add to cart
 
MS1494R1
View Documents
Thermo Scientific™
MS1494R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Thermo Scientific™ Mesothelioma Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: HBME-1

Isotype: IgM/κ

Species Reactivity: Human. Others not known.

Immunogen: A suspension of human mesotheliema cells from patients with malignant epithelial mesothelioma

Positive Control: Mesothelioma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Mesothelioma Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
HBME-1
Monoclonal
Murine
Cancer and Tumor Biology
A suspension of human mesotheliema cells from patients with malignant epithelial mesothelioma
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit